Product Details

SNP ID
rs11585055
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:159808058 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAAGGACTGGAGACAGTGACACC[A/G]TACCAGTGCCATCCAAGGGCCTCCA
Phenotype
MIM: 613562
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FCRL6 PubMed Links

Gene Details

Gene
FCRL6
Gene Name
Fc receptor like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004310.2 Intron NP_001004310.2
NM_001284217.1 Intron NP_001271146.1
XM_005245128.3 Intron XP_005245185.1
XM_005245129.3 Intron XP_005245186.1
XM_005245131.3 Intron XP_005245188.1
XM_006711292.3 Intron XP_006711355.1
XM_011509480.1 Intron XP_011507782.1
XM_011509481.2 Intron XP_011507783.1
XM_017001176.1 Intron XP_016856665.1
XM_017001177.1 Intron XP_016856666.1

View Full Product Details