Product Details

SNP ID
rs4503258
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71103387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCCCTTCCCCCAGTGCCCTTTG[C/T]ATCGTCCCCTCCCCCACCCCTGCTG
Phenotype
MIM: 300033 MIM: 308380
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXorf65 PubMed Links

Gene Details

Gene
CXorf65
Gene Name
chromosome X open reading frame 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025265.2 3036 Intron NP_001020436.1
XM_005262244.3 3036 Intron XP_005262301.1
Gene
FOXO4
Gene Name
forkhead box O4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170931.1 3036 UTR 3 NP_001164402.1
NM_005938.3 3036 UTR 3 NP_005929.2
XM_011530957.2 3036 UTR 3 XP_011529259.1
Gene
IL2RG
Gene Name
interleukin 2 receptor subunit gamma
There are no transcripts associated with this gene.

View Full Product Details