Product Details

SNP ID
rs12461291
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49161260 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAATTCTATAGCCAATTGTGTGC[G/T]TGTGTTTGTGTGTGTGTGTGTTTAG
Phenotype
MIM: 142705 MIM: 606936
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HRC PubMed Links

Gene Details

Gene
HRC
Gene Name
histidine rich calcium binding protein
There are no transcripts associated with this gene.

Gene
TRPM4
Gene Name
transient receptor potential cation channel subfamily M member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195227.1 Intron NP_001182156.1
NM_001321281.1 Intron NP_001308210.1
NM_001321282.1 Intron NP_001308211.1
NM_001321283.1 Intron NP_001308212.1
NM_001321285.1 Intron NP_001308214.1
NM_017636.3 Intron NP_060106.2
XM_005259017.1 Intron XP_005259074.1

View Full Product Details