Product Details

SNP ID
rs4596815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103253613 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTGGGGCCAGATTTTAATATTA[G/T]GCAAAAATGAATTCAAGGCCTAAAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TCEAL8 PubMed Links

Gene Details

Gene
TCEAL8
Gene Name
transcription elongation factor A like 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006684.1 521 UTR 3 NP_001006685.1
NM_153333.2 521 UTR 3 NP_699164.1

View Full Product Details