Product Details
- SNP ID
-
rs361355
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:142463509 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTCTCAGGTCTCCAGTTCCCTAAT[G/T]ATAGCTCTGAGCTGAATGTGAACGC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TRBV5-4
PubMed Links
Gene Details
- Gene
- TRBV5-4
- Gene Name
- T cell receptor beta variable 5-4
There are no transcripts associated with this gene.
View Full Product Details