Product Details

SNP ID
rs2353532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:135711355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTGAAACACAGGCAAATATTAA[C/T]ACAATAGGCTAGGAAGCTATAATAC
Phenotype
MIM: 300648
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CT45A1 PubMed Links

Gene Details

Gene
CT45A1
Gene Name
cancer/testis antigen family 45, member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017417.2 Intron NP_001017417.1
XM_005278141.3 Intron XP_005278198.1
XM_011531352.1 Intron XP_011529654.1

View Full Product Details