Product Details

SNP ID
rs4845480
Assay Type
Functionally tested
NCBI dbSNP Submissions
57
Location
Chr.1:152759960 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTGTCCTACGTGCAGTGCGAAGC[C/G]TCACAACCTGTTCAGACTTGCTTCG
Phenotype
MIM: 613260
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KPRP PubMed Links

Gene Details

Gene
KPRP
Gene Name
keratinocyte proline rich protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025231.1 471 Silent Mutation GCC,GCG A124A NP_001020402.1
XM_011509571.1 471 Silent Mutation GCC,GCG A124A XP_011507873.1

View Full Product Details