Product Details

SNP ID
rs4076879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32999311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAAACGTCTCCTTCCCTGTTAGC[A/G]TGACACATGCCACCCTGTGGACCCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RHPN2 PubMed Links

Gene Details

Gene
RHPN2
Gene Name
rhophilin Rho GTPase binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033103.4 Intron NP_149094.3

View Full Product Details