Product Details
- SNP ID
-
hCV26698939
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:32995998 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTTCCGCGGCACAGGCACCCCCA[C/T]AGAGGGAACACAGTCTAGGCTACTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RHPN2
PubMed Links
Gene Details
- Gene
- RHPN2
- Gene Name
- rhophilin Rho GTPase binding protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033103.4 |
|
Intron |
|
|
NP_149094.3 |
View Full Product Details