Product Details

SNP ID
rs1133145
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53163492 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAATTTTAAAAGGCAGAGTCTTT[C/T]TTCTTCAGGAGCTTGGAGAGAAATA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZNF347 PubMed Links

Gene Details

Gene
ZNF347
Gene Name
zinc finger protein 347
There are no transcripts associated with this gene.

Gene
ZNF665
Gene Name
zinc finger protein 665
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024733.3 3098 UTR 3 NP_079009.3
XM_005259266.4 3098 Intron XP_005259323.1
XM_011527325.2 3098 Intron XP_011525627.1
XM_011527327.2 3098 Intron XP_011525629.1

View Full Product Details