Product Details

SNP ID
rs7152848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:70470404 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAACAGTTCTACAGCAACGTGGCC[C/T]GGCCTGCTAGCAGAAAATAGTACTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAM20P1 PubMed Links

Gene Details

Gene
ADAM20P1
Gene Name
ADAM metallopeptidase domain 20 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details