Product Details

SNP ID
rs7918487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:23193706 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTTCTCTTCTCACCTGTCCAGGG[C/T]CCCCCTCCCCCAGCGACCCTGATTA
Phenotype
MIM: 607194
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf67 PubMed Links

Gene Details

Gene
C10orf67
Gene Name
chromosome 10 open reading frame 67
There are no transcripts associated with this gene.

Gene
PTF1A
Gene Name
pancreas specific transcription factor, 1a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178161.2 787 Missense Mutation CCC,TCC P263S NP_835455.1

View Full Product Details