Product Details
- SNP ID
-
hCV26742446
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:42792947 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGCTGAAGCTGGCTTTTGGGTTC[C/T]GTCTTCCAGGGGTAGGAGATTTTGC
- Phenotype
-
MIM: 611448
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BMS1
PubMed Links
Gene Details
- Gene
- BMS1
- Gene Name
- BMS1, ribosome biogenesis factor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014753.3 |
|
Intron |
|
|
NP_055568.3 |
View Full Product Details