Product Details

SNP ID
rs2997636
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:162806305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTATCGCAGGTAAGTCAAGTAAC[C/T]GGCATTGATCTTGGCCTCTGTTAGT
Phenotype
MIM: 606756
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSD17B7 PubMed Links

Gene Details

Gene
HSD17B7
Gene Name
hydroxysteroid 17-beta dehydrogenase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304512.1 Intron NP_001291441.1
NM_001304513.1 Intron NP_001291442.1
NM_016371.3 Intron NP_057455.1

View Full Product Details