Product Details

SNP ID
rs2581467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:99581150 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGTGAACATTTCTTTCCTTAGAG[A/T]TAGTAAGTAGTCTGTCAGTATTTTC
Phenotype
MIM: 600660
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MEF2A PubMed Links

Gene Details

Gene
MEF2A
Gene Name
myocyte enhancer factor 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130926.2 Intron NP_001124398.1
NM_001130927.2 Intron NP_001124399.1
NM_001130928.2 Intron NP_001124400.1
NM_001171894.2 Intron NP_001165365.1
NM_001319206.1 Intron NP_001306135.1
NM_005587.3 Intron NP_005578.2
XM_005254914.2 Intron XP_005254971.1
XM_005254915.2 Intron XP_005254972.1
XM_005254916.3 Intron XP_005254973.1
XM_011521571.2 Intron XP_011519873.1
XM_011521572.2 Intron XP_011519874.1
XM_011521573.2 Intron XP_011519875.1
XM_011521576.2 Intron XP_011519878.1
XM_011521577.2 Intron XP_011519879.1
XM_011521578.2 Intron XP_011519880.1
XM_011521579.2 Intron XP_011519881.1
XM_011521581.2 Intron XP_011519883.1
XM_011521582.2 Intron XP_011519884.1
XM_011521583.2 Intron XP_011519885.1
XM_011521585.2 Intron XP_011519887.1
XM_011521586.2 Intron XP_011519888.1
XM_011521587.2 Intron XP_011519889.1
XM_011521590.2 Intron XP_011519892.1
XM_017022190.1 Intron XP_016877679.1
XM_017022191.1 Intron XP_016877680.1
XM_017022192.1 Intron XP_016877681.1
XM_017022193.1 Intron XP_016877682.1
XM_017022194.1 Intron XP_016877683.1
XM_017022195.1 Intron XP_016877684.1
XM_017022196.1 Intron XP_016877685.1
XM_017022197.1 Intron XP_016877686.1
XM_017022198.1 Intron XP_016877687.1
XM_017022199.1 Intron XP_016877688.1
XM_017022200.1 Intron XP_016877689.1
XM_017022201.1 Intron XP_016877690.1
XM_017022202.1 Intron XP_016877691.1
XM_017022203.1 Intron XP_016877692.1
XM_017022204.1 Intron XP_016877693.1

View Full Product Details