Product Details

SNP ID
rs35367128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49533284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCGGGAGGTGGGGCGGTGACTCA[C/G]GCAGGCCTGGAGAGGGCTCTGCTGT
Phenotype
MIM: 601437
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FCGRT PubMed Links

Gene Details

Gene
FCGRT
Gene Name
Fc fragment of IgG receptor and transporter
There are no transcripts associated with this gene.

Gene
RCN3
Gene Name
reticulocalbin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020650.2 Intron NP_065701.2
XM_005259089.3 Intron XP_005259146.1
XM_011527143.1 Intron XP_011525445.1
XM_017027023.1 Intron XP_016882512.1

View Full Product Details