Product Details

SNP ID
rs10857884
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:111469517 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCCAAAATCTCAGTAGCTCAATAC[A/G]GAAAAGTTTACTTTTTGCTCATATA
Phenotype
MIM: 603270
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP5F1 PubMed Links

Gene Details

Gene
ATP5F1
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1
There are no transcripts associated with this gene.

Gene
C1orf162
Gene Name
chromosome 1 open reading frame 162
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300834.1 Intron NP_001287763.1
NM_001300835.1 Intron NP_001287764.1
NM_174896.3 Intron NP_777556.1
XM_011540712.2 Intron XP_011539014.1
XM_017000323.1 Intron XP_016855812.1
XM_017000324.1 Intron XP_016855813.1

View Full Product Details