Product Details

SNP ID
rs12029516
Assay Type
Functionally tested
NCBI dbSNP Submissions
49
Location
Chr.1:111341391 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAATCTTTTAATCATCTGAATG[C/T]GTCAAAGTATTGAGAAAATATCAGA
Phenotype
MIM: 614234
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIFO PubMed Links

Gene Details

Gene
PIFO
Gene Name
primary cilia formation
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300831.1 Intron NP_001287760.1
NM_181643.5 Intron NP_857594.2
XM_005270472.1 Intron XP_005270529.1
XM_017000322.1 Intron XP_016855811.1

View Full Product Details