Product Details

SNP ID
rs2696923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:49032049 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAAGATCGATTTCTGTAAAGGAT[A/G]CATAAAATTCCTGTGGGTCTATTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM49B PubMed Links

Gene Details

Gene
TRIM49B
Gene Name
tripartite motif containing 49B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206626.1 Intron NP_001193555.1
XM_017017561.1 Intron XP_016873050.1

View Full Product Details