Product Details
- SNP ID
-
rs12042573
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
36
- Location
-
Chr.1:234757649 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTAAAAACCTTATTTATTTTAATA[C/T]AAGAGAGGAGTTTCCTTGTTTCCGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PP2672
PubMed Links
Gene Details
- Gene
- PP2672
- Gene Name
- uncharacterized LOC100130249
There are no transcripts associated with this gene.
View Full Product Details