Product Details

SNP ID
rs1049150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54818467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACATTCCCATCTTCCATGGCAGA[A/T]TATTCCAGGAGAGCTTCAACATGAG
Phenotype
MIM: 604945 MIM: 604946
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KIR2DL4 PubMed Links

Gene Details

Gene
KIR2DL4
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 4
There are no transcripts associated with this gene.

Gene
KIR3DL1
Gene Name
killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322168.1 286 Silent Mutation ATA,TTA I75L NP_001309097.1
NM_013289.2 286 Missense Mutation ATA,TTA I75L NP_037421.2
Gene
LOC102724300
Gene Name
uncharacterized LOC102724300
There are no transcripts associated with this gene.

View Full Product Details