Product Details

SNP ID
rs605241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72135728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCACAGAGCAAGCTGGTGTCAGA[A/G]CCTGGACCTACAGCGCTGTTGGTGG
Phenotype
MIM: 602469
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOLR3 PubMed Links

Gene Details

Gene
FOLR3
Gene Name
folate receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000804.3 19 UTR 5 NP_000795.2
NM_001318045.1 19 UTR 5 NP_001304974.1
XM_011544876.1 19 Intron XP_011543178.1

View Full Product Details