Product Details
- SNP ID
-
rs2916410
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:4192656 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTTGCCCCTCCTCCAGGGTCCTCC[A/G]TCTCCAACATAGTATCACCATCTCC
- Phenotype
-
MIM: 607806
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OTOP1
PubMed Links
Gene Details
- Gene
- OTOP1
- Gene Name
- otopetrin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_177998.1 |
|
Intron |
|
|
NP_819056.1 |
View Full Product Details