Product Details
- SNP ID
-
hCV26934095
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:4238356 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGTTATCACTTATAAGCTGCTATT[A/C]TATCTTTCATTGAGAAAAACACTAT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM128
PubMed Links
Gene Details
- Gene
- TMEM128
- Gene Name
- transmembrane protein 128
View Full Product Details