Product Details

SNP ID
rs111387149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:4237268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCAACAGTCACTGAATATAAGC[G/T]GACACAAATTAATATAATTTCAACA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMEM128 PubMed Links

Gene Details

Gene
TMEM128
Gene Name
transmembrane protein 128
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297551.1 Intron NP_001284480.1
NM_001297552.1 Intron NP_001284481.1
NM_032927.3 Intron NP_116316.1
XM_005248034.2 Intron XP_005248091.1

View Full Product Details