Product Details

SNP ID
rs9810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:112819410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATATCCAAAATGAAATAATTCT[C/T]AGGAGTACCCATACAGTATTAGAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BMT2 PubMed Links

Gene Details

Gene
BMT2
Gene Name
base methyltransferase of 25S rRNA 2 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152556.2 3679 UTR 3 NP_689769.2

View Full Product Details