Product Details

SNP ID
rs8081537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:79734466 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGACAGTGATGAGGCAGACACTG[C/T]GTATCTGGGAACATTTTTTTTTTTT
Phenotype
MIM: 616997
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENPP7 PubMed Links

Gene Details

Gene
ENPP7
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178543.4 Intron NP_848638.3
XM_011524737.1 Intron XP_011523039.1

View Full Product Details