Product Details

SNP ID
hCV26962208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:206065675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAGAGGATAGGGACAGGAAAGT[C/A]CTGCCTGTGGGACAACAGGGATCAG
Phenotype
MIM: 616088
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
C1orf186 PubMed Links

Gene Details

Gene
C1orf186
Gene Name
chromosome 1 open reading frame 186
There are no transcripts associated with this gene.

Gene
LOC100505650
Gene Name
uncharacterized LOC100505650
There are no transcripts associated with this gene.

View Full Product Details