Product Details

SNP ID
rs11588867
Assay Type
Functionally tested
NCBI dbSNP Submissions
27
Location
Chr.1:212373693 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCAATTTTAAAATGGGCAAGAGAT[C/T]TGAATAGAGATTTATCCCAAAAGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM206 PubMed Links

Gene Details

Gene
TMEM206
Gene Name
transmembrane protein 206
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198862.1 Intron NP_001185791.1
NM_018252.2 Intron NP_060722.2
XM_005273177.3 Intron XP_005273234.1
XM_011509717.2 Intron XP_011508019.1
XM_011509718.2 Intron XP_011508020.1
XM_017001634.1 Intron XP_016857123.1
XM_017001635.1 Intron XP_016857124.1
XM_017001636.1 Intron XP_016857125.1
XM_017001637.1 Intron XP_016857126.1

View Full Product Details