Product Details
- SNP ID
-
rs2680712
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:58432504 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGGTGAAAAACATTTCAGGCAGAGA[C/T]AATAGCAAGTGTAAAGGCTGGAGCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HSF5
PubMed Links
Gene Details
- Gene
- HSF5
- Gene Name
- heat shock transcription factor 5
View Full Product Details