Product Details

SNP ID
rs6452709
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:86289797 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTGCTGTAGGCCCTGCCCCTTT[A/G]CAAGCAAGCTCTGTCCTGCTATAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NBPF22P PubMed Links

Gene Details

Gene
NBPF22P
Gene Name
neuroblastoma breakpoint family member 22, pseudogene
There are no transcripts associated with this gene.

View Full Product Details