Product Details

SNP ID
rs2516408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31495714 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGGAGGGTTCATAGTCTAATGGG[A/G]GAGGAGGGTAGAAGATGGTAGGTGA
Phenotype
MIM: 602436
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102725068 PubMed Links

Gene Details

Gene
LOC102725068
Gene Name
uncharacterized LOC102725068
There are no transcripts associated with this gene.

Gene
MICB
Gene Name
MHC class I polypeptide-related sequence B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289160.1 Intron NP_001276089.1
NM_001289161.1 Intron NP_001276090.1
NM_005931.4 Intron NP_005922.2

View Full Product Details