Product Details

SNP ID
rs2696059
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:184653973 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGATGCTCAACCCAGCTGTTAAA[A/T]TTTTCAAGCTGTGCAGATATGAAGG
Phenotype
MIM: 600636 MIM: 615421
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
CASP3 PubMed Links

Gene Details

Gene
CASP3
Gene Name
caspase 3
There are no transcripts associated with this gene.

Gene
PRIMPOL
Gene Name
primase and DNA directed polymerase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300767.1 Intron NP_001287696.1
NM_001300768.1 Intron NP_001287697.1
NM_152683.3 Intron NP_689896.1
XM_011531719.1 Intron XP_011530021.1
XM_011531720.1 Intron XP_011530022.1
XM_011531721.2 Intron XP_011530023.1
XM_011531723.1 Intron XP_011530025.1
XM_011531724.2 Intron XP_011530026.1
XM_011531725.1 Intron XP_011530027.1
XM_011531726.2 Intron XP_011530028.1
XM_011531729.1 Intron XP_011530031.1
XM_011531730.2 Intron XP_011530032.1
XM_017007864.1 Intron XP_016863353.1
XM_017007865.1 Intron XP_016863354.1
XM_017007866.1 Intron XP_016863355.1
XM_017007867.1 Intron XP_016863356.1
XM_017007868.1 Intron XP_016863357.1
XM_017007869.1 Intron XP_016863358.1
XM_017007870.1 Intron XP_016863359.1
XM_017007871.1 Intron XP_016863360.1
XM_017007872.1 Intron XP_016863361.1
XM_017007873.1 Intron XP_016863362.1
XM_017007874.1 Intron XP_016863363.1
XM_017007875.1 Intron XP_016863364.1

View Full Product Details