Product Details

SNP ID
hCV27058686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49304996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATTAGCAGAAATTAATTTTTGTG[C/A]CAGTATTGTTGCATTAGTATGGAAA
Phenotype
MIM: 300737
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
GAGE10 PubMed Links

Gene Details

Gene
GAGE10
Gene Name
G antigen 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098413.3 Intron NP_001091883.3
Gene
PPP1R3F
Gene Name
protein phosphatase 1 regulatory subunit 3F
There are no transcripts associated with this gene.

View Full Product Details