Product Details
- SNP ID
-
hCV27058739
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:49313056 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTAGTGTTTGTTTACACCTTCTC[A/T]GGGTAGGCTGAGGCCTTCCCCACCA
- Phenotype
-
MIM: 300737
MIM: 300733
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GAGE10
PubMed Links
Gene Details
- Gene
- GAGE10
- Gene Name
- G antigen 10
- Gene
- GAGE12J
- Gene Name
- G antigen 12J
There are no transcripts associated with this gene.
View Full Product Details