Product Details
- SNP ID
-
rs1006371
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
45
- Location
-
Chr.1:15154957 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGTAAGTCAGCGGGTGGAAGGCTT[G/T]CCCCTCTAGGACCGGCCGGCTTCTG
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C1orf195
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79668780] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C1orf195
- Gene Name
- chromosome 1 open reading frame 195
There are no transcripts associated with this gene.
- Gene
- TMEM51
- Gene Name
- transmembrane protein 51
- Gene
- TMEM51-AS1
- Gene Name
- TMEM51 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details