Product Details

SNP ID
rs1048745
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55939063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAACATTCTTCATGACACTATTT[A/G]TTATGAGGAAAGTTGCAGCTAAATA
Phenotype
MIM: 600694
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL6ST PubMed Links

Gene Details

Gene
IL6ST
Gene Name
interleukin 6 signal transducer
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190981.1 5089 UTR 3 NP_001177910.1
NM_002184.3 5089 UTR 3 NP_002175.2
NM_175767.2 5089 UTR 3 NP_786943.1
XM_017009441.1 5089 UTR 3 XP_016864930.1
XM_017009442.1 5089 Intron XP_016864931.1

View Full Product Details