Product Details
- SNP ID
-
rs11960951
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:6348844 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAACCACTTGTACTTCAGGGAAAC[A/C]CCCATCAAGAACCTCATTGATGGCT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LY86-AS1
PubMed Links
Gene Details
- Gene
- LY86-AS1
- Gene Name
- LY86 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details