Product Details

SNP ID
rs3925948
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:18131660 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAACATAGAGCATATAGCATATA[C/T]ACAGAGCATATTTACAGAGATTAGT
Phenotype
MIM: 608072 MIM: 187680
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NHLRC1 PubMed Links
Additional Information
For this assay, SNP(s) [rs114611199] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
NHLRC1
Gene Name
NHL repeat containing E3 ubiquitin protein ligase 1
There are no transcripts associated with this gene.

Gene
TPMT
Gene Name
thiopurine S-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000367.3 Intron NP_000358.1
XM_011514839.2 Intron XP_011513141.1
XM_017011241.1 Intron XP_016866730.1

View Full Product Details