Product Details

SNP ID
rs2413057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31682086 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCATTGATTATGAGACGGAATTC[C/G]AGCCTGGAGACGGAGCGACGGCCGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRR14L PubMed Links

Gene Details

Gene
PRR14L
Gene Name
proline rich 14 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173566.2 10650 UTR 3 NP_775837.2
XM_006724219.3 10650 UTR 3 XP_006724282.1
XM_006724220.3 10650 UTR 3 XP_006724283.1
XM_006724221.2 10650 UTR 3 XP_006724284.1
XM_011530080.2 10650 UTR 3 XP_011528382.1
XM_011530081.2 10650 UTR 3 XP_011528383.1
XM_017028736.1 10650 UTR 3 XP_016884225.1
XM_017028737.1 10650 UTR 3 XP_016884226.1

View Full Product Details