Product Details

SNP ID
rs12289616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125071413 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGATCAGAGTCCCTGCTGCAGC[A/G]TGAAGGCTGTCTTCATTCTGCCTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC37A2 PubMed Links

Gene Details

Gene
SLC37A2
Gene Name
solute carrier family 37 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145290.1 Intron NP_001138762.1
NM_198277.2 Intron NP_938018.1

View Full Product Details