Product Details

SNP ID
rs3828364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:87229139 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTTTAATCGCCTAATACCTGTCA[A/G]ATATTTTCAAAGACATAAATCTCAG
Phenotype
MIM: 609512
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHMP2B PubMed Links

Gene Details

Gene
CHMP2B
Gene Name
charged multivesicular body protein 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244644.1 Intron NP_001231573.1
NM_014043.3 Intron NP_054762.2
XM_011533576.2 Intron XP_011531878.1
Gene
MIR4795
Gene Name
microRNA 4795
There are no transcripts associated with this gene.

View Full Product Details