Product Details

SNP ID
rs4994685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:43917196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCGGGAGGGGAGAGGTCACTGTA[C/T]TCATTTAGGGGCTAGAGGTCTTTTG
Phenotype
MIM: 611163
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TOX2 PubMed Links

Gene Details

Gene
TOX2
Gene Name
TOX high mobility group box family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098796.1 Intron NP_001092266.1
NM_001098797.1 Intron NP_001092267.1
NM_001098798.1 Intron NP_001092268.1
NM_032883.2 Intron NP_116272.1
XM_006723884.1 Intron XP_006723947.1
XM_017028109.1 Intron XP_016883598.1

View Full Product Details