Product Details

SNP ID
rs2284379
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34006836 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGCACACTGGTGTTTGCACAGGGA[C/T]GAAATCGCCTAATGATGCATTTCTC
Phenotype
MIM: 614663
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RALY PubMed Links

Gene Details

Gene
RALY
Gene Name
RALY heterogeneous nuclear ribonucleoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007367.3 Intron NP_031393.2
NM_016732.2 Intron NP_057951.1
XM_005260334.4 Intron XP_005260391.1
XM_005260336.4 Intron XP_005260393.1
XM_011528694.2 Intron XP_011526996.1
XM_011528695.2 Intron XP_011526997.1
XM_017027731.1 Intron XP_016883220.1

View Full Product Details