Product Details

SNP ID
rs35544677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100429331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGCGCCAGTTTCAGCACGTGCT[A/G]CCTCTACCCGCGCAAGCCCAAAAGG
Phenotype
MIM: 611478
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MEPCE PubMed Links

Gene Details

Gene
MEPCE
Gene Name
methylphosphate capping enzyme
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001194990.1 542 Intron NP_001181919.1
NM_001194991.1 542 Intron NP_001181920.1
NM_001194992.1 542 Intron NP_001181921.1
NM_019606.5 542 Intron NP_062552.2
XM_011516410.2 542 UTR 5 XP_011514712.1
Gene
PPP1R35
Gene Name
protein phosphatase 1 regulatory subunit 35
There are no transcripts associated with this gene.

Gene
ZCWPW1
Gene Name
zinc finger CW-type and PWWP domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258008.1 542 Intron NP_001244937.1
NM_017984.4 542 Intron NP_060454.3
XM_005250479.2 542 Intron XP_005250536.1
XM_005250480.2 542 Intron XP_005250537.1
XM_006716035.3 542 Intron XP_006716098.1
XM_006716036.2 542 Intron XP_006716099.1
XM_006716037.3 542 Intron XP_006716100.1
XM_006716038.3 542 Intron XP_006716101.1
XM_006716040.3 542 Intron XP_006716103.1
XM_017012379.1 542 Intron XP_016867868.1
XM_017012380.1 542 Intron XP_016867869.1

View Full Product Details