Product Details
- SNP ID
-
rs2928220
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:21370144 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGCTTTTTAAACATCTATTGAAAT[A/G]ACTTTGTGGTTTTTATCTTTAATTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF738
PubMed Links
Gene Details
- Gene
- ZNF738
- Gene Name
- zinc finger protein 738
There are no transcripts associated with this gene.
View Full Product Details