Product Details

SNP ID
rs2505306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:27508596 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATTGTTACCTATGTTCCAAGTAA[C/G]TAAATATAGTCAAGATGTTAAGTAG
Phenotype
MIM: 602207
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RAB18 PubMed Links

Gene Details

Gene
RAB18
Gene Name
RAB18, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256410.1 Intron NP_001243339.1
NM_001256411.1 Intron NP_001243340.1
NM_001256412.1 Intron NP_001243341.1
NM_001256415.1 Intron NP_001243344.1
NM_021252.4 Intron NP_067075.1

View Full Product Details