Product Details

SNP ID
rs997723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:57088071 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGAGGTAGTAGATCCTGTGGGCAT[C/G]TTGTCCCATTGAAATCATCTCATAT
Phenotype
MIM: 602449
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AKAP1 PubMed Links

Gene Details

Gene
AKAP1
Gene Name
A-kinase anchoring protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242902.1 Intron NP_001229831.1
NM_001242903.1 Intron NP_001229832.1
NM_003488.3 Intron NP_003479.1
XM_005257707.2 Intron XP_005257764.1
XM_005257709.2 Intron XP_005257766.1
XM_017025189.1 Intron XP_016880678.1
XM_017025190.1 Intron XP_016880679.1
XM_017025191.1 Intron XP_016880680.1

View Full Product Details