Product Details

SNP ID
hCV27305414
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:46853177 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATATCTTTGTAATACTTCCTCTG[C/T]AGATACATTCACTTAGTTCAAACCT
Phenotype
MIM: 600388
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ADGRF5 PubMed Links

Gene Details

Gene
ADGRF5
Gene Name
adhesion G protein-coupled receptor F5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098518.1 5085 UTR 3 NP_001091988.1
NM_015234.4 5085 UTR 3 NP_056049.4
Gene
MEP1A
Gene Name
meprin A subunit alpha
There are no transcripts associated with this gene.

View Full Product Details