Product Details

SNP ID
hCV27305422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:46855886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACATTTCAGGTAAATGCCATGA[G/T]CGGGGTGAAAGAGTAGGGGTGTTGA
Phenotype
MIM: 600388
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADGRF5 PubMed Links

Gene Details

Gene
ADGRF5
Gene Name
adhesion G protein-coupled receptor F5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098518.1 Intron NP_001091988.1
NM_015234.4 Intron NP_056049.4
Gene
MEP1A
Gene Name
meprin A subunit alpha
There are no transcripts associated with this gene.

View Full Product Details